Learn More
Cayman Chemical ANTIBODY 4EGI-1 5mg
SDP
Supplier:  Cayman Chemical 153625 MG
Encompass_Preferred
An inhibitor of the initiation of translation that blocks the interaction of eIF4G with eIF4E (KD = 25 μM); prevents the expression of oncogenic proteins in mammalian cancer cells, leading to apoptosis; blocks translation in some viruses that do not require eIF4E or a cap structure for initiation
Catalog No. 50-297-2179