Learn More
Cayman Chemical ANTIBODY 5 S-HpETE 250ug
SDP
Supplier:  Cayman Chemical 44230250 UG
Encompass_Preferred
A product of 5-LO catalyzed oxidation of arachidonic acid; further metabolism by 5-LO gives LTA4, a key intermediate in the formation of the 4-series LTs
Catalog No. 50-294-5274