Learn More
Cayman Chemical ANTIBODY 5R 6S-EET 50UG
SDP
Supplier:  Cayman Chemical 900434350 UG
Encompass_Preferred
An oxylipin; depolarizes transepithelial voltage in isolated rabbit renal cortical collecting duct when applied to the luminal side at 1 µM but to a lesser extent than 5S(6R)-EET; degrades in solution into 5,6-DiHET and 5(6)-δ-lactone, which can be converted to 5(6)-DiHET and quantified by GC-MS
Catalog No. 50-289-5063