Learn More
Cayman Chemical ANTIBODY 6-epI COTC 1mg
SDP
Supplier:  Cayman Chemical 377461 MG
Encompass_Preferred
A derivative of COTC with anticancer activity; cytotoxic to A549 and H460 NSCLC cells (IC50s = 170 and 158 µM, respectively)
Catalog No. 50-291-6844