Learn More
Cayman Chemical ANTIBODY 77PD-Q 500ug
SDP
Supplier:  Cayman Chemical 40927500 UG
Encompass_Preferred
A PPD-Q and derivative of 77PD; toxic to V. fischeri (EC50 = 13.6 mg/L); has been found in PM2.5 samples collected at the roadside in China
Catalog No. 50-294-4882