Learn More
Cayman Chemical ANTIBODY 8 S-HETrE 250ug
SDP
Supplier:  Cayman Chemical 36360250 UG
Encompass_Preferred
A metabolite of GLA; serum levels are decreased in a mouse model of high-fat high-sucrose diet-induced obesity
Catalog No. 50-291-2104