Learn More
Cayman Chemical ANTIBODY 852A 10mg
SDP
Supplier:  Cayman Chemical 3951210 MG
Encompass_Preferred
A TLR7 agonist; selectively induces NF-κB signaling in HEK293 cells expressing recombinant human TLR7 but not HEK293 cells expressing recombinant human TLR8 or TLR9 at 1 µM; induces IFN-α production in plasmacytoid dendritic cell-enriched isolated human PBMCs from 10-1,000 ng/ml; delays the onset of lung metastasis in a B16/F10 murine melanoma model at 150 mg/kg
Catalog No. 50-291-8906