Learn More
Cayman Chemical ANTIBODY 9-OxoDE 50ug
SDP
Supplier:  Cayman Chemical 3842050 UG
Encompass_Preferred
9-OxoODE results from oxidation of the allylic hydroxyl of either 9(S)- or 9(R)-HODE. Rabbit reticulocyte plasma and mitochondrial membranes contain both 9- and 13-oxoODEs, representing about 2% of the total linoleate residues in the membranes. Most of these oxidized linoleate residues are esterified to membrane lipids.{2262,2326,2261}
Catalog No. 50-291-6743