Learn More
Cayman Chemical ANTIBODY 9 R-PAHSA 1mg
SDP
Supplier:  Cayman Chemical 180221 MG
Encompass_Preferred
A stereoisomer of 9-PAHSA; the predominant form of 9-PAHSA that accumulates in adipose tissue in vivo; cell lines favor the production of 9(R)-PAHSA
Catalog No. 50-296-6330