Learn More
Cayman Chemical ANTIBODY A17 100mg
SDP
Supplier:  Cayman Chemical 39686100 MG
Encompass_Preferred
A bile acid derivative; reduces oleic acid-induced triglyceride accumulation, increases cell viability, and decreases oleic acid-induced lipid peroxidation in primary rat hepatocytes at 10 and 25 µM; reduces levels of CD36 in primary rat hepatocytes at 25 µM; reduces serum triglyceride, total cholesterol, and LDL levels, reverses hepatic steatosis, and decreases hepatic triglyceride, but not total cholesterol, levels in a hamster model of high-fat diet-induced NASH at 50 mg/kg
Catalog No. 50-291-9272