Learn More
Cayman Chemical ANTIBODY A286982 25mg
SDP
Supplier:  Cayman Chemical 2764625 MG
Encompass_Preferred
An inhibitor that blocks the integrin-ligand interaction between LFA-1 and ICAM-1 (IC50 = 35 nM); inhibits adhesion of JY-8 cells to ICAM-1-coated microtiter plates (IC50 = 44 nM)
Catalog No. 50-294-0687