Learn More
Cayman Chemical ANTIBODY ABD-F 10mg
SDP
Supplier:  Cayman Chemical 3889210 MG
Encompass_Preferred
A thiol-reactive fluorogenic reagent; has been used to detect and quantify thiol-containing amino acids, protein and tissue thiols, and disulfide linkages; ex/em = 389/513 nm, respectively
Catalog No. 50-291-8705