Learn More
Cayman Chemical ANTIBODY ABT-639 1mg
SDP
Supplier:  Cayman Chemical 287421 MG
Encompass_Preferred
A T-type calcium channel blocker; inhibits inactivated-state Cav3.2 channels (IC50 = 2.3 μM) and inactivated-state Cav3.1 and Cav3.3 channels by 50 and 39%, respectively, at 10 μM; selectively inhibits inactivated-state T-type calcium currents over resting-state currents in rat DRG neurons (IC50s = 7.6 and >30 μM, respectively); reverses the hind limb grip force deficit in a rat model of MIA-induced osteoarthritic pain (ED50 = 2 mg/kg); dose-dependently increases the paw withdrawal threshold in rat SNL and CCI models of neuropathic pain; attenuates cold allodynia in the CCI rat model at ≥3 mg/kg
Catalog No. 50-293-9846