Learn More
Cayman Chemical ANTIBODY Ac-PAL-AMC 1mg
SDP
Supplier:  Cayman Chemical 265921 MG
Encompass_Preferred
A fluorogenic substrate for the 20S proteasome; AMC is released upon enzymatic cleavage by the β1i/LMP2 subunit of the immunoproteasome and its fluorescence can be used to quantify enzyme activity; selective for the immunoproteasome over the constitutive proteasome; 351/430 nm, respectively
Catalog No. 50-293-2664