Learn More
Cayman Chemical ANTIBODY Acephate 50mg
SDP
Supplier:  Cayman Chemical 2403850 MG
Encompass_Preferred
An organophosphate insecticide active against a variety of insects; inhibits acetylcholinesterase in the house fly with a topical 24-hour LD50 value of 1.8 UG/g; induces reversible hyperglycemia after two and six hours and increases liver glycogen levels by 3.5-fold after six hours at 140 mg/kg
Catalog No. 50-293-4154