Learn More
Cayman Chemical ANTIBODY ActInonIn 25mg
SDP
Supplier:  Cayman Chemical 1660525 MG
Encompass_Preferred
A peptidomimetic antibiotic produced by Actinomyces that inhibits aminopeptidases, including MMP-1 (Ki = 300 nM), MMP-3 (Ki = 1,700 nM), MMP-8 (Ki = 130 nM), and MMP-9 (Ki = 330 nM); also targets peptide deformylase and meprin α (Ki = 20 nM)
Catalog No. 50-296-5690