Learn More
Cayman Chemical ANTIBODY AdefovIr 10mg
SDP
Supplier:  Cayman Chemical 1865010 MG
Encompass_Preferred
An active metabolite of adefovir dipivoxil; formed from adefovir dipivoxil by CES2; inhibits cytopathogenicity induced by HSV-1 and HSV-2 (EC50 = 7 UG/ml for both in E6SM cells), HIV-1 and HIV-2 (EC50s = 1.8 and 2.5 UG/ml, respectively, in CEM cells), and VZV (EC50 = 10 UG/ml in human embryonic lung fibroblasts); reduces increases in lymph node viral DNA and serum IgG levels in a mouse model of AIDS induced by infection with LP-BM5 at 50 mg/kg; inhibits tumor growth induced by MSV infection in mice at 50 mg/kg
Catalog No. 50-298-0202