Learn More
Cayman Chemical ANTIBODY AG-1024 1mg
SDP
Supplier:  Cayman Chemical 148331 MG
Encompass_Preferred
A selective inhibitor of IGF-1R that inhibits insulin-stimulated cellular proliferation and IGF-1R autophosphorylation with IC50 values of 0.4 and 7 μM, respectively; inhibits proliferation of human breast cancer MCF-7 cells (10 nM) and human prostate cancer DU145 cells (IC50 = 2.5 μM)
Catalog No. 50-297-1939