Learn More
Cayman Chemical ANTIBODY AG-120 25mg
SDP
Supplier:  Cayman Chemical 1989425 MG
Encompass_Preferred
An orally available inhibitor of mutant IDH1 that may induce cellular differentiation as well as inhibit proliferation in IDH1-expressing tumor cells
Catalog No. 50-298-1175