Learn More
Cayman Chemical ANTIBODY Akt-I-1 5mg
SDP
Supplier:  Cayman Chemical 410165 MG
Encompass_Preferred
An Akt1 inhibitor (IC50 = 4.6 µM); selective for Akt1 over Akt2, Akt3, and SGK (IC50s = >250 µM for all)
Catalog No. 50-294-4905