Learn More
Cayman Chemical ANTIBODY ALK-IN-1 5mg
SDP
Supplier:  Cayman Chemical 187755 MG
Encompass_Preferred
A potent inhibitor of ALK (IC50 = 0.07 nM) that is less effective at IGF-1R and InsR (IC50s = 3.2 and 100 nM, respectively); drives cell death in (ALK+) Karpas-299 lymphoma cells with an IC50 value of 41.5 nM, while killing (ALK-) U937 cells with an IC50 value of 1,718 nM
Catalog No. 50-296-6216