Learn More
Cayman Chemical ANTIBODY AM6545 5mg
SDP
Supplier:  Cayman Chemical 163165 MG
Encompass_Preferred
A novel CB1 selective neutral antagonist with low CNS penetration (Ki = 1.7 and 523 nM for CB1 and CB2, respectively); reduces food intake, food-reinforced behavior and weight gain; improves metabolic profiles of mice with diet-induced obesity
Catalog No. 50-296-8913