Learn More
Cayman Chemical ANTIBODY AMG-TIe2-1 10mg
SDP
Supplier:  Cayman Chemical 2885010 MG
Encompass_Preferred
An inhibitor of Tie2 and VEGFR2 (IC50s = 1 and 3 nM, respectively, in a HTRF assay); inhibits angiopoietin 1-induced Tie2 autophosphorylation in EA.hy926 human endothelial cells (IC50 = 10 nM).
Catalog No. 50-294-0052