Learn More
Cayman Chemical ANTIBODY AmrInon 1g
SDP
Supplier:  Cayman Chemical 238981 G
Encompass_Preferred
An inhibitor of PDE3 (IC50 = 19.5 µM); increases the rate and force of heart contraction in anesthetized and unanesthetized dogs
Catalog No. 50-299-4370