Learn More
Cayman Chemical ANTIBODY AP39 10mg
SDP
Supplier:  Cayman Chemical 1710010 MG
Encompass_Preferred
A compound used to increase the levels of H2S within mitochondria; stimulates mitochondrial electron transport and improves cellular bioenergetic function at lower concentrations (30-100 nM), while having an inhibitory effect at 300 nM; is effective in vivo
Catalog No. 50-296-6905