Learn More
Cayman Chemical ANTIBODY ApremIlast 25mg
SDP
Supplier:  Cayman Chemical 1850225 MG
Encompass_Preferred
An orally active inhibitor of PDE4 with an IC50 value of 74 nM when tested in U937 cell lysates with 1 mM cAMP; decreases epidermal thickness in a mouse model of psoriasis, reduces both TNF-α and MMP-3 production in gut cells from patients with IBD, and improves clinical score in mouse models of arthritis
Catalog No. 50-296-5254