Learn More
Cayman Chemical ANTIBODY APY-29 10mg
SDP
Supplier:  Cayman Chemical 2291310 MG
Encompass_Preferred
A modulator of IRE1α function; binds to the ATP-binding site on IRE1α, where it inhibits autophosphorylation (IC50 = 280 nM) and enhances its RNase function (EC50 = 460 nM); restores the ability of dephosphoylated IRE1α to cleave XBP1 mRNA in a dose-dependent manner; enhances IRE1α oligomerization in a crosslinking assay
Numéro de catalogue. 50-299-3971
Ajouter au panier
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.