Learn More
Cayman Chemical ANTIBODY AR-7 5mg
SDP
Supplier:  Cayman Chemical 229465 MG
Encompass_Preferred
A RARα antagonist; increases chaperone-mediated autophagy in an RARα-dependent manner, increases LAMP-2A expression, and inhibits paraquat-induced decreases in cell viability in mouse fibroblasts at 20 μM; selectively cytotoxic to HepG2 cells grown under hypoxic over normoxic conditions (IC50s = 10 and >1,000 μM, respectively)
Catalog No. 50-299-5007