Learn More
Cayman Chemical ANTIBODY AR231453 5mg
SDP
Supplier:  Cayman Chemical 348985 MG
Encompass_Preferred
An agonist of GPR119; increases cAMP accumulation in HIT-T15 β cells that endogenously express high levels of GPR119 (EC50 = 4.7 nM); increases insulin release from HIT-T15 cells (EC50 = 3.5 nM); reduces plasma glucose levels in wild-type, KK/Ay, and db/db diabetic mice at 20 mg/kg
Catalog No. 50-291-1035