Learn More
Cayman Chemical ANTIBODY ARQ 092 10mg
SDP
Supplier:  Cayman Chemical 2138810 MG
Encompass_Preferred
A selective, allosteric, orally bioavailable, pan-Akt inhibitor (IC50s = 5.0, 4.5, and 16 nM for Akt1, Akt2, and Akt3, respectively)
Catalog No. 50-299-4079