Learn More
Raybiotech Inc C-Series Human Periodontal Disease Antibody Array 1 Kit.
Supplier:  Raybiotech Inc AAHPDD12
Encompass_Preferred
Detects 20 human periodontal disease-associated cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-2, IL-4, IL-6, IL-8 (CXCL8), IL-10, IL-12 p70, IFN-gamma, TGF beta 1, TNF alpha, Osteoprotegerin (TNFRSF11B), IL-17A, MIP-1 alpha (CCL3), MMP-13, MMP-9, Osteopontin (SPP1), RANK (TNFRSF11A), CRP (C-Reactive Protein), Osteoactivin (GPNMB)
Catalog No. 50-194-8632