Learn More
Raybiotech Inc C-Series Human EGFR Phosphorylation Antibody Array 1 Kit.
Supplier:  Raybiotech Inc AAHPER14
Encompass_Preferred
Detects 17 site specific and pan Human EGFRs. Suitable for all liquid sample types but intended for use with cell and tissue lysates. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: EGFR (P-Tyr845), EGFR (P-Tyr992), EGFR (P-Tyr1045), EGFR (P-Tyr1068), EGFR (P-Tyr1086), EGFR (P-Tyr1148), EGFR (P-Tyr1173), EGFR (P-Ser1046/1047), EGFR (P-Ser1070), ErbB2 (P-Tyr877), ErbB2 (P-Tyr1112), ErbB2 (P-Tyr 1221/1222), ErbB2 (P-Tyr1248), ErbB2 (P-Thr686), ErbB2 (P-Ser1113), ErbB3 (P-Tyr1289), ErbB4 (P-Tyr1284)
Catalog No. 50-194-8636