Learn More
Raybiotech Inc G-Series Human Angiogenesis Array 1000 Kit.
Supplier:  Raybiotech Inc GSHANG10001
Encompass_Preferred
A combination of 2 non-overlapping arrays to measure the relative expression levels of 60 human angiogenic proteins. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. View protein list on RayBiotech’s website – GSH-ANG-1000-1
Catalog No. 50-195-0356