Learn More
Raybiotech Inc G-Series Human Immune Checkpoint Molecule Array 1 Kit.
Supplier:  Raybiotech Inc GSHICM11
Encompass_Preferred
Detects 10 Human Immune Checkpoint Molecules. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: B7-1 (CD80), B7-2 (CD86), B7-H1 (PD-L1), B7-H2 (ICOS L), B7-H3 (CD276), CD28 (Tp44), CTLA-4 (CD152), ICOS (CD278), PD-1 (CD279), PD-L2 (B7-DC)
Catalog No. 50-195-0400