Learn More
Raybiotech Inc Quantibody Human Bone Metabolism Array 1000 Kit.
Supplier:  Raybiotech Inc QAHBMA10001
Encompass_Preferred
Detects 41 Human Cytokines. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: Activin A, aFGF (FGF-1), Amphiregulin, bFGF, BMP-4, BMP-9, E-Selectin, ICAM-1 (CD54), IGF-1, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-6, IL-8 (CXCL8), IL-11, IL-17A, MCP-1 (CCL2), M-CSF, MIP-1 alpha (CCL3), MMP-2, MMP-9, MMP-13, Osteoactivin (GPNMB), P-Cadherin, RANK (TNFRSF11A), SDF-1 alpha (CXCL12 alpha), Sonic Hedgehog N-Terminal (Shh-N), TGF beta 1, TGF beta 2, TNF alpha, VCAM-1 (CD106)
Catalog No. 50-195-0797