Learn More
Cayman Chemical ANTIBODY ARS853 500ug
SDP
Supplier:  Cayman Chemical 19137500 UG
Encompass_Preferred
An inhibitor of K-RASG12C; decreases the level of active GTP-bound K-RAS by 95% (10 µM); inhibits proliferation preferentially in lung cancer cell lines containing the K-RASG12C mutation, including H358 cells (IC50 = 2.5 µM)
Catalog No. 50-298-3948