Learn More
Cayman Chemical ANTIBODY ARV-766 1mg
SDP
Supplier:  Cayman Chemical 419851 MG
Encompass_Preferred
A PROTAC that drives androgen receptor degradation (DC50 = <1 nM); inhibits tumor growth in various LNCaP and VCaP prostate cancer mouse xenograft models, including an enzalutamide-insensitive, non-castrated VCaP model
Catalog No. 50-294-3916