Learn More
Cayman Chemical ANTIBODY ASP3026 5mg
SDP
Supplier:  Cayman Chemical 184915 MG
Encompass_Preferred
An ALK inhibitor (IC50s = 3.5, 10, and 5.4 nM for the wild-type, ALKF1174L, and ALKR1275Q enzymes, respectively); selective for ALK over a panel of 86 kinases at 1,000 nM but also inhibits ROS1 and TNK2 (IC50s = 8.9 and 5.8 nM, respectively); inhibits the proliferation of Ba/F3 cells expressing NPM-ALK or ELM4-ALK fusion proteins (IC50s = 84 and 56 nM, respectively), as well as several crizotinib-resistant Ba/F3 cells expressing NPM-ALK or ELM4-ALK mutants; induces tumor regression in an NCI H2228 NSCLC mouse xenograft model at 10 and 30 mg/kg
Catalog No. 50-296-6885