Learn More
Cayman Chemical ANTIBODY Aspalaton 10mg
SDP
Supplier:  Cayman Chemical 1364410 MG
Encompass_Preferred
An anti-platelet aggregator (IC50 = 180 µM, in vitro) that prolongs bleeding time significantly in a rodent model of thromboembolism; at 24 mg/kg, generates antioxidant and neuroprotective effects against kainic acid-induced epilepsy in rat hippocampus
Catalog No. 50-295-3594