Learn More
Cayman Chemical ANTIBODY ATB-343 50mg
SDP
Supplier:  Cayman Chemical 1304550 MG
Encompass_Preferred
A hybrid molecule of an H2S donor and the NSAID indomethacin that does not cause gastric damage or promote leukocyte adherence to vascular endothelium
Catalog No. 50-295-3125