Learn More
Cayman Chemical ANTIBODY ATX-001 1mg
SDP
Supplier:  Cayman Chemical 390371 MG
Encompass_Preferred
An ionizable cationic lipid; has been used in combination with other lipids in the formation of LNPs for the delivery of mRNA in vivo
Catalog No. 50-291-7227