Learn More
Cayman Chemical ANTIBODY AVE-0991 10mg
SDP
Supplier:  Cayman Chemical 3783710 MG
Encompass_Preferred
A Mas receptor agonist; binds to the Mas receptor in bovine aortic endothelial cell membranes and COS cells (IC50s = 21 and 47.5 nM, respectively); inhibits perivascular inflammation and increases plaque stability in an ApoE-/- mouse model of atherosclerosis at 0.58 µmol/kg per day; inhibits the formation of angiotensin II-induced abdominal aortic aneurysm in ApoE-/- mice
Catalog No. 50-291-5919