Learn More
Cayman Chemical ANTIBODY AZ 3146 25mg
SDP
Supplier:  Cayman Chemical 1999125 MG
Encompass_Preferred
A selective Mps1 inhibitor (IC50 = 35 nM); inhibits the recruitment of Mad1, Mad2, and CENP-E to kinetochores
Catalog No. 50-298-1499