Learn More
Cayman Chemical ANTIBODY AZ 505 10mg
SDP
Supplier:  Cayman Chemical 1687510 MG
Encompass_Preferred
An inhibitor of SMYD2 (IC50 = 0.12 μM); selective for SMYD2 over SMYD3, DOT1L, EZH2, GLP, G9A, and SET7/9 (IC50s = >83.3 μM); delays cyst growth in EArly- and late-stage Pdk1 conditional knockout mouse models of polycystic kidney disease at 10 mg/kg
Catalog No. 50-297-9854