Learn More
Cayman Chemical ANTIBODY AZD 1656 1mg
SDP
Supplier:  Cayman Chemical 418391 MG
Encompass_Preferred
An HK4 activator; activates HK4 and induces glucose phosphorylation in a cell-free assay (EC50 = 61 nM); decreases blood glucose levels in an OGTT in rats at 0.3 mg/kg; reduces blood glucose levels and increases plasma insulin levels in mice when administered at doses of 2, 4.5, or 9 mg/kg
Catalog No. 50-294-2757