Learn More
Cayman Chemical ANTIBODY AZD 3965 10mg
SDP
Supplier:  Cayman Chemical 1991210 MG
Encompass_Preferred
A selective inhibitor of MCT1 (Ki = 1.6 nM), killing tumor cells that are reliant on glycolysis by blocking lactate transport; increases intratumor lactate levels and decreases tumor growth in mice bearing SCLC xenografts
Catalog No. 50-298-3176