Learn More
Cayman Chemical ANTIBODY AZD 5462 10mg
SDP
Supplier:  Cayman Chemical 4095610 MG
Encompass_Preferred
An RXFP1 agonist; induces cAMP mobilization in HEK293 cells expressing the human RXFP1 (EC50 = 15.85 nM); increases heart rate and MABP in rats; increases the left ventricular ejection fraction (LVEF) in a cynomolgus monkey model of heart failure induced by a high-fat diet and age at 1 or 10 mg/kg per day
Catalog No. 50-294-2858