Learn More
Cayman Chemical ANTIBODY AZD 8835 10mg
SDP
Supplier:  Cayman Chemical 2916610 MG
Encompass_Preferred
An inhibitor of PI3Kα and PI3Kδ (IC50s = 6.2 and 5.7 nM, respectively); selective for PI3Kα and PI3Kδ over PI3Kβ and PI3Kγ (IC50s = 431 and 90 nM, respectively); inhibits Akt phosphorylation in PIK3CA constitutively active mutant BT474 cells and Jeko-1 B cells that express endogenous PI3Kδ (IC50s = 57 and 49 nM, respectively); reduces tumor growth and Akt phosphorylation in an SKOV3 mouse xenograft model at 25 mg/kg; increases the activity of CD8+ effector T cells and reduces tumor growth in a C26 mouse syngeneic tumor model
Catalog No. 50-293-9979