Learn More
Cayman Chemical ANTIBODY-Baclofen 25g
SDP
Supplier:  Cayman Chemical 1860025 G
Encompass_Preferred
A GABAB receptor agonist (IC50 = 180 nM); induces norepinephrine release from isolated rat atria (IC50 = 4.5 μM); increases muscle rigidity in spastic mice at 10 mg/kg; inhibits the norepinephrine-induced group II flexor reflex in anesthetized rats; reduces cocaine-induced hyperlocomotion in rats and binge-like ethanol intake in mice
Catalog No. 50-296-6230