Learn More
Cayman Chemical ANTIBODY BAMEA-O16B 5mg
SDP
Supplier:  Cayman Chemical 374395 MG
Encompass_Preferred
An ionizable cationic lipid; has been used in the generation of LNPs for the delivery of CRISPR complementary sgRNA and Cas9 mRNA for genome editing in vitro and in vivo; LNPs containing BAMEA-O16B encapsulating Cas9 mRNA and sgRNAs targeting PCSK9 accumulate in the liver and decrease serum PCSK9 levels in mice
Catalog No. 50-291-6237